Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191927.1 | NP_001178856.1 | 6 | 1163 | ||
XM_006248342.2 | XP_006248404.1 | 6 | 1215 | |||
XM_006248343.2 | XP_006248405.1 | 6 | 1212 |