Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109055.1 | NP_001102525.1 | 3 | 1352 | ||
XM_006248459.2 | XP_006248521.1 | 3 | 1455 | |||
XM_006248460.2 | XP_006248522.1 | 3 | 1256 | |||
XM_006248461.2 | XP_006248523.1 | 3 | 1090 |