Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025760.1 | NP_001020931.1 | 6 | 1673 | ||
XM_006249015.2 | XP_006249077.1 | 4 | 1497 | |||
GenBank mRNA | BC099191.1 | - | 6 | 1673 | 7383463 |