Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pm20d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109068.1 | NP_001102538.1 | 8, 9 | 1012 | ||
GenBank mRNA | FQ210945.1 | - | 4, 5 | 299 |