Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261834.1 | NP_001248763.1 | 9 | 1779 | ||
NM_004749.3 | NP_004740.2 | 9 | 1823 | |||
NM_030900.3 | NP_112162.1 | 7 | 1493 | |||
NM_199122.2 | NP_954573.1 | 7 | 1472 | |||
GenBank mRNA | AB023165.1 | - | 7 | 1464 | ||
AF011792.1 | - | 7 | 1158 | |||
AK001281.1 | - | 7 | 1441 | |||
AK022575.1 | - | 9 | 1759 | |||
AK092132.1 | - | 7 | 2957 | |||
AK222943.1 | - | 7 | 1429 | |||
AK298155.1 | - | 7 | 1172 | |||
AK300489.1 | - | 9 | 1738 | |||
AL833840.1 | - | 9 | 1792 | |||
BC002732.2 | - | 7 | 1262 | |||
BC014918.1 | - | 9 | 1753 | 3906670 | ||
BC017235.2 | - | 9 | 1750 | 3538504 |