Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170459.1 | NP_001163930.1 | 20, 21 | 3613 | ||
XM_006251459.1 | XP_006251521.1 | 22, 23 | 3999 | |||
XM_006251460.1 | XP_006251522.1 | 22, 23 | 3999 | |||
XM_006251462.2 | XP_006251524.1 | 22, 23 | 4381 | |||
XM_006251463.2 | XP_006251525.1 | 22, 23 | 3838 | |||
XM_006251464.2 | XP_006251526.1 | 22, 23 | 3930 | |||
XM_006251466.2 | XP_006251528.1 | 20, 21 | 3990 | |||
XM_006251467.2 | XP_006251529.1 | 22, 23 | 4227 | |||
XM_008770334.1 | XP_008768556.1 | 22, 23 | 5207 | |||
XM_008770335.1 | XP_008768557.1 | 23, 24 | 4399 | |||
XM_008770336.1 | XP_008768558.1 | 19, 20 | 5243 | |||
XM_008770337.1 | XP_008768559.1 | 20, 21 | 9085 | |||
XM_008770338.1 | XP_008768560.1 | 22, 23 | 3991 | |||
GenBank mRNA | AB510354.1 | - | 20, 21 | 3623 |