Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024903.1 | NP_001020074.2 | 2 | 291 | ||
XM_006251526.2 | XP_006251588.1 | 3 | 554 | |||
XM_006251527.2 | XP_006251589.1 | 2 | 1676 | |||
GenBank mRNA | BC098047.1 | - | 2 | 291 | 7457109 |