Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109097.1 | NP_001102567.1 | 2 | 134 | ||
XM_006251655.2 | XP_006251717.1 | 2 | 181 | |||
GenBank mRNA | FQ214748.1 | - | 2 | 134 | ||
FQ214808.1 | - | 2 | 134 | |||
FQ214853.1 | - | 2 | 117 | |||
FQ215077.1 | - | 136 | ||||
FQ215190.1 | - | 2 | 136 | |||
FQ215360.1 | - | 2 | 140 | |||
FQ215382.1 | - | 2 | 136 | |||
FQ215728.1 | - | 2 | 136 | |||
FQ215901.1 | - | 2 | 138 | |||
FQ215918.1 | - | 2 | 134 | |||
FQ216019.1 | - | 2 | 174 | |||
FQ216101.1 | - | 2 | 136 | |||
FQ216115.1 | - | 2 | 135 | |||
FQ216248.1 | - | 2 | 135 | |||
FQ216396.1 | - | 2 | 118 | |||
FQ216753.1 | - | 2 | 118 | |||
FQ217380.1 | - | 156 | ||||
FQ224894.1 | - | 2 | 138 | |||
FQ224977.1 | - | 2 | 134 |