Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109104.1 | NP_001102574.1 | 1 | 242 | ||
XM_008770884.1 | XP_008769106.1 | 2 | 169 | |||
XM_008770885.1 | XP_008769107.1 | 1 | 288 |