Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110490.2 | NP_001103960.1 | 7 | 795 | ||
NM_001277158.1 | NP_001264087.1 | 8 | 810 | |||
NM_001277159.1 | NP_001264088.1 | 8 | 800 | |||
NM_001277165.1 | NP_001264094.1 | 6 | 926 | |||
NM_001277166.1 | NP_001264095.1 | 7 | 785 | |||
XM_006252637.2 | XP_006252699.1 | 6 | 982 | |||
XM_006252640.2 | XP_006252702.1 | 7 | 841 | |||
XM_006252641.2 | XP_006252703.1 | 6 | 982 | |||
XM_006252644.2 | XP_006252706.1 | 7 | 841 | |||
XM_006252645.2 | XP_006252707.1 | 6 | 982 | |||
XM_006252647.1 | XP_006252709.1 | 7 | 841 | |||
GenBank mRNA | BC083721.1 | - | 6 | 733 | 7190925 | |
FM122871.1 | - | 5 | 551 | |||
FQ214774.1 | - | 6 | 639 | |||
FQ216348.1 | - | 7 | 790 | |||
FQ217970.1 | - | 8 | 806 |