Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024904.1 | NP_001020075.1 | 2 | 1125 | ||
XM_006243832.1 | XP_006243894.1 | 2 | 1154 | |||
XM_006243835.2 | XP_006243897.1 | 2 | 1194 | |||
GenBank mRNA | BC098020.1 | - | 2 | 1125 | 7455232 |