Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080208.1 | NP_001073677.1 | 2 | 317 | ||
GenBank mRNA | EF078967.1 | - | 2 | 317 |