Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1562608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134607.1 | NP_001128079.1 | 8 | 1384 | ||
XM_003751756.3 | XP_003751804.2 | 10 | 1784 |