Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem170a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134608.1 | NP_001128080.1 | 2 | 285 | ||
XM_008772528.1 | XP_008770750.1 | 2 | 285 |