Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138549.1 | NP_612558.1 | 5, 6 | 413 | ||
XM_006255216.2 | XP_006255278.1 | 4, 5 | 548 | |||
XM_006255217.2 | XP_006255279.1 | 3, 4 | 415 | |||
XM_006255218.2 | XP_006255280.1 | 4, 5 | 373 | |||
GenBank mRNA | BC059115.1 | - | 5, 6 | 376 | 6920522 | |
EU000473.1 | - | 5, 6 | 376 | |||
FQ212009.1 | - | 4, 5 | 354 | |||
FQ215654.1 | - | 4, 5 | 470 | |||
FQ215778.1 | - | 3, 4 | 280 | |||
FQ216428.1 | - | 3, 4 | 420 | |||
FQ219100.1 | - | 5, 6 | 353 | |||
FQ220726.1 | - | 5, 6 | 362 | |||
S45663.1 | - | 5, 6 | 413 |
Gene Symbol: RGD1560015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_574311.6 | XP_574311.1 | 1 | 314 |