Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127655.1 | NP_001121127.1 | 2 | 198 | ||
XM_006229782.2 | XP_006229844.1 | 2 | 616 | |||
XM_006229783.2 | XP_006229845.1 | 2 | 206 | |||
GenBank mRNA | BC159412.1 | - | 2 | 198 | 8367388 |