Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam111a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109163.1 | NP_001102633.1 | 3 | 708 | ||
XM_006231125.1 | XP_006231187.1 | 3 | 738 | |||
GenBank mRNA | FQ226103.1 | - | 3 | 707 | ||
FQ230484.1 | - | 3 | 706 |