Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109166.1 | NP_001102636.1 | 1 | 746 | ||
XM_006231554.2 | XP_006231616.1 | 1 | 707 |