Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145456.1 | NP_001138928.1 | 2 | 272 | ||
XM_006256385.2 | XP_006256447.1 | 2 | 2174 | |||
XM_006256386.2 | XP_006256448.1 | 2 | 2174 | |||
XM_006256387.2 | XP_006256449.1 | 2 | 2171 | |||
XM_006256388.2 | XP_006256450.1 | 2 | 2038 | |||
XM_006256389.2 | XP_006256451.1 | 2 | 265 | |||
XM_006256390.2 | XP_006256452.1 | 2 | 1845 | |||
XM_006256391.2 | XP_006256453.1 | 2 | 1844 | |||
GenBank mRNA | FJ640984.1 | - | 2 | 272 | ||
FJ640985.1 | - | 2 | 351 | |||
FQ222665.1 | - | 2 | 360 |