Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SOX2 SIRNA Assays ›
Gene Symbol:
Sox2
Gene Name:
SRY (sex determining region Y)-box 2
Gene Aliases:
RGD1565646
Chromosome Location:
Chr.2: 121062030 - 121203880 on Build Rnor_6.0
UniGene ID:
Rn.17369
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s178243
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sox2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001109181.1
NP_001102651.1
1
740
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011443, AK149358, AK131933, AB221649, U31967, and others.
Panther Classification:
Molecular Function -
HMG box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
osteoblast differentiation
eye development
cell fate specification
endodermal cell fate specification
positive regulation of neuroblast proliferation
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell cycle arrest
sensory perception of sound
response to wounding
response to organic substance
regulation of gene expression
stem cell population maintenance
forebrain neuron differentiation
pituitary gland development
adenohypophysis development
cerebral cortex development
positive regulation of cell-cell adhesion
negative regulation of Wnt signaling pathway
male genitalia development
positive regulation of epithelial cell differentiation
forebrain development
olfactory placode formation
response to retinoic acid
somatic stem cell population maintenance
tissue regeneration
inner ear morphogenesis
regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAPK cascade
tongue development
cell fate commitment
negative regulation of cell differentiation
positive regulation of cell differentiation
negative regulation of neuron differentiation
positive regulation of neuron differentiation
negative regulation of osteoblast differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
pigment biosynthetic process
lung alveolus development
embryonic organ development
anatomical structure formation involved in morphogenesis
neuron fate commitment
inner ear development
diencephalon morphogenesis
stem cell differentiation
negative regulation of epithelial cell proliferation
regulation of neurogenesis
detection of mechanical stimulus involved in sensory perception of sound
detection of mechanical stimulus involved in equilibrioception
retina morphogenesis in camera-type eye
lens induction in camera-type eye
epithelial tube branching involved in lung morphogenesis
response to growth factor
negative regulation of canonical Wnt signaling pathway
neuronal stem cell population maintenance
transcription regulatory region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
miRNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0