Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100794.1 | NP_001094264.1 | 1 | 195 | ||
GenBank mRNA | BC086515.1 | - | 1 | 164 | ||
BC166888.1 | - | 1 | 195 | 6891440 | ||
BC166920.1 | - | 1 | 195 | 6919256 | ||
DY564389.1 | - | 1 | 174 | |||
FQ212650.1 | - | 1 | 174 | |||
FQ220191.1 | - | 1 | 174 | |||
FQ226064.1 | - | 1 | 181 | |||
FQ227364.1 | - | 1 | 181 |