Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079893.2 | NP_001073362.1 | 2 | 1001 | ||
NM_001270835.1 | NP_001257764.1 | 2 | 988 | |||
NM_001270836.1 | NP_001257765.1 | 2 | 992 | |||
GenBank mRNA | BC158555.1 | - | 2 | 970 | 7311099 | |
FQ213995.1 | - | 2 | 989 |
Gene Symbol: Dusp14l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749277.3 | XP_003749325.1 | 1 | 1027 |