Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004776.3 | NP_004767.1 | 9 | 1307 | ||
GenBank mRNA | AB004550.1 | - | 9 | 1225 | ||
AB871482.2 | - | 9 | 1106 | |||
AF038663.1 | - | 9 | 1123 | |||
BC074821.2 | - | 9 | 1146 | 30915537 | ||
BC074873.2 | - | 9 | 1146 | 30915234 | ||
BC104987.1 | - | 9 | 1146 | 8143990 | ||
BC112265.1 | - | 9 | 1146 | 8327733 |