Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177688.1 | NP_001171159.1 | 10 | 1216 | ||
XM_006234347.2 | XP_006234409.1 | 10 | 1288 | |||
GenBank mRNA | BC160919.1 | - | 10 | 1216 | 7458373 |