Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004781.3 | NP_004772.1 | 3 | 331 | ||
XM_017002841.1 | XP_016858330.1 | 3 | 325 | |||
GenBank mRNA | AK316558.1 | - | 3 | 323 | ||
BC003570.1 | - | 3 | 251 | 3544610 | ||
BC005941.1 | - | 3 | 331 | 4069433 | ||
BC007050.1 | - | 3 | 267 | 3681914 | ||
BT007327.1 | - | 2 | 216 | |||
CR542145.1 | - | 2 | 216 | |||
CR542154.1 | - | 2 | 216 | |||
U64520.1 | - | 3 | 240 |