Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191972.1 | NP_001178901.1 | 2 | 620 |
Gene Symbol: NEWGENE_1565788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762952.1 | XP_008761174.1 | 2 | 631 |