Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280558.1 | NP_001267487.1 | 16 | 1812 | ||
NM_006032.3 | NP_006023.1 | 15 | 1800 | |||
XM_005268216.1 | XP_005268273.1 | 16 | 1756 | |||
XM_005268217.1 | XP_005268274.1 | 14 | 1450 | |||
GenBank mRNA | AB009288.1 | - | 15 | 1600 | ||
AK125195.1 | - | 14 | 2362 | |||
AK222675.1 | - | 15 | 1560 | |||
AK293655.1 | - | 16 | 1812 | |||
AK314182.1 | - | 15 | 1613 | |||
AL050397.1 | - | 3 | 1463 | |||
AL137688.1 | - | 7 | 4128 | |||
BC018046.1 | - | 15 | 1606 | 4799650 |