Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001690.3 | NP_001681.2 | 8 | 1014 | ||
GenBank mRNA | AF113129.1 | - | 8 | 941 | ||
AK293804.1 | - | 9 | 1048 | |||
AK295150.1 | - | 3 | 198 | |||
AK314779.1 | - | 8 | 977 | |||
BC013138.1 | - | 8 | 962 | 4454722 | ||
BT006672.1 | - | 7 | 906 | |||
L09235.1 | - | 7 | 921 |