Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17922
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
7
2151
NM_001272020.1
NP_001258949.1
6
2685
NM_004796.5
NP_004787.2
17
3514
NM_138970.4
NP_620426.2
6
2070
NR_073546.1
8
3463
NR_073547.1
21
6497
XM_005268218.2
XP_005268275.1
23
5878
XM_006720322.2
XP_006720385.1
22
5875
XM_006720323.2
XP_006720386.1
21
5539
XM_011537363.1
XP_011535665.1
23
5887
XM_011537364.2
XP_011535666.1
24
5849
XM_011537365.2
XP_011535667.1
23
5675
XM_011537366.1
XP_011535668.1
22
5863
XM_011537367.1
XP_011535669.1
22
5797
XM_011537368.1
XP_011535670.1
23
5584
XM_011537369.1
XP_011535671.1
23
5575
XM_011537370.1
XP_011535672.1
22
5551
XM_011537371.1
XP_011535673.1
23
5263
XM_011537372.1
XP_011535674.1
23
5254
XM_011537373.1
XP_011535675.1
22
5164
XM_011537377.2
XP_011535679.1
19
4179
XM_017021790.1
XP_016877279.1
24
5987
XM_017021791.1
XP_016877280.1
24
5770
XM_017021792.1
XP_016877281.1
22
5670
XM_017021793.1
XP_016877282.1
22
5860
XM_017021794.1
XP_016877283.1
21
5851
XM_017021795.1
XP_016877284.1
21
5646
XM_017021796.1
XP_016877285.1
21
5827
XM_017021797.1
XP_016877286.1
20
5827
XM_017021798.1
XP_016877287.1
20
5824
XM_017021799.1
XP_016877288.1
22
5788
XM_017021800.1
XP_016877289.1
22
5485
XM_017021801.1
XP_016877290.1
21
5458
XM_017021802.1
XP_016877291.1
20
5479
XM_017021803.1
XP_016877292.1
22
5233
XM_017021804.1
XP_016877293.1
22
5227
XM_017021805.1
XP_016877294.1
21
5218
XM_017021806.1
XP_016877295.1
21
5212
XM_017021807.1
XP_016877296.1
22
5173
GenBank mRNA
AB018286.1
-
17
3495
AJ316284.2
-
21
6285
AJ493127.1
-
8
2539
AK056530.1
-
6
2685
AK096247.1
-
1
676
AL833739.1
-
6
1365
BC059368.1
-
6
1996
BC068469.1
-
6
1996
BC142649.1
-
7
1584
BC150194.1
-
7
1584
BC152457.1
-
17
3495
8862590
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053817, XM_008764731, XM_008764733, XM_008764732, L14851.This siRNA also targets the following transcripts in Mouse: NM_172544, NM_001198587, NM_001252074, AK034255, AK129205, and others.
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline