Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17924
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
6
1969
NM_001272020.1
NP_001258949.1
5
1879
NM_004796.5
NP_004787.2
16
3332
NM_138970.4
NP_620426.2
5
1879
NR_073546.1
6
1969
NR_073547.1
19
5003
XM_005268218.2
XP_005268275.1
22
5072
XM_006720322.2
XP_006720385.1
21
5060
XM_006720323.2
XP_006720386.1
20
5036
XM_011537363.1
XP_011535665.1
22
5072
XM_011537364.2
XP_011535666.1
23
5034
XM_011537365.2
XP_011535667.1
22
4860
XM_011537366.1
XP_011535668.1
21
5048
XM_011537367.1
XP_011535669.1
21
4982
XM_011537368.1
XP_011535670.1
22
5072
XM_011537369.1
XP_011535671.1
22
5072
XM_011537370.1
XP_011535672.1
21
5048
XM_011537371.1
XP_011535673.1
22
5072
XM_011537372.1
XP_011535674.1
22
5072
XM_011537373.1
XP_011535675.1
21
4982
XM_011537377.2
XP_011535679.1
18
3364
XM_017021790.1
XP_016877279.1
23
5172
XM_017021791.1
XP_016877280.1
23
4955
XM_017021792.1
XP_016877281.1
21
4855
XM_017021793.1
XP_016877282.1
21
5045
XM_017021794.1
XP_016877283.1
20
5036
XM_017021795.1
XP_016877284.1
20
4831
XM_017021796.1
XP_016877285.1
20
5021
XM_017021797.1
XP_016877286.1
19
5012
XM_017021798.1
XP_016877287.1
19
5009
XM_017021799.1
XP_016877288.1
21
4982
XM_017021800.1
XP_016877289.1
21
4982
XM_017021801.1
XP_016877290.1
20
4955
XM_017021802.1
XP_016877291.1
19
4976
XM_017021803.1
XP_016877292.1
21
5051
XM_017021804.1
XP_016877293.1
21
5045
XM_017021805.1
XP_016877294.1
20
5036
XM_017021806.1
XP_016877295.1
20
5021
XM_017021807.1
XP_016877296.1
21
4982
GenBank mRNA
AB018286.1
-
16
3313
AJ316284.2
-
19
4791
AJ493127.1
-
6
1045
AK056530.1
-
5
1879
AL833739.1
-
5
1174
BC059368.1
-
5
1805
BC068469.1
-
5
1805
BC142649.1
-
6
1402
BC150194.1
-
6
1402
BC152457.1
-
16
3313
8862590
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172544, NM_001198587, NM_001252074, AK129205, BC060719, and others.
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline