Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191980.1 | NP_001178909.1 | 8 | 1056 | ||
XM_008763419.1 | XP_008761641.1 | 8 | 2399 | |||
XM_008763420.1 | XP_008761642.1 | 7 | 964 | |||
XM_008763421.1 | XP_008761643.1 | 6 | 1008 |