Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025053.2 | NP_001020224.2 | 1 | 664 | ||
XM_006241297.2 | XP_006241359.1 | 1 | 663 | |||
GenBank mRNA | AB040675.1 | - | 1 | 418 | ||
BC105818.1 | - | 1 | 664 | 7368805 |