Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191988.1 | NP_001178917.1 | 5 | 474 | ||
GenBank mRNA | DQ630444.1 | - | 5 | 453 |