Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191989.1 | NP_001178918.1 | 5 | 1214 | ||
XM_006241492.2 | XP_006241554.1 | 5 | 2252 | |||
XM_006241493.2 | XP_006241555.1 | 5 | 2094 | |||
XM_006241494.2 | XP_006241556.1 | 5 | 2270 | |||
XM_006241495.2 | XP_006241557.1 | 5 | 2249 |