Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: RGD1563692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109311.1 | NP_001102781.1 | 6 | 1586 | ||
XM_006245547.2 | XP_006245609.1 | 5 | 2613 | |||
GenBank mRNA | BC162036.1 | - | 5 | 1399 | 7097280 |