Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109673.1 | NP_001103143.1 | 3, 4 | 299 | ||
XM_006244336.2 | XP_006244398.1 | 3, 4 | 363 | |||
GenBank mRNA | BC129098.1 | - | 3, 4 | 314 | 8373484 | |
BC169032.1 | - | 3, 4 | 299 | 9091720 |