Gene Symbol: Bex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037365.1 | NP_001032442.1 | 567 | |||
GenBank mRNA | AY833554.1 | - | 567 |
Gene Symbol: LOC100912195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229316.2 | XP_006229378.1 | 3 | 625 |