Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771344.1 | XP_008769566.1 | 348 | |||
GenBank mRNA | FQ215711.1 | - | 343 |
Gene Symbol: LOC100912478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253887.2 | XP_006253949.1 | 2 | 348 |
Gene Symbol: LOC103689964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253890.2 | XP_006253952.1 | 348 |