Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1561667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748704.3 | XP_003748752.1 | 2 | 398 | ||
XM_008758801.1 | XP_008757023.1 | 1 | 360 |
Gene Symbol: LOC685793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001065293.4 | XP_001065293.2 | 2 | 354 |
Gene Symbol: RGD1561185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758870.1 | XP_008757092.1 | 1 | 382 |