Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172088.1 | NP_001165559.1 | 6 | 711 |
Gene Symbol: LOC497796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111310.1 | NP_001104780.1 | 867 | |||
XM_006237126.2 | XP_006237188.1 | 7 | 917 | |||
GenBank mRNA | BC093399.1 | - | 867 | 7389533 |
Gene Symbol: LOC502907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763434.1 | XP_008761656.1 | 7 | 1390 |