Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025066.1 | NP_001020237.1 | 3 | 625 | ||
XM_008765642.1 | XP_008763864.1 | 2 | 470 | |||
XM_008765643.1 | XP_008763865.1 | 3 | 1275 | |||
GenBank mRNA | BC083768.1 | - | 3 | 625 | 7193209 |