Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192018.1 | NP_001178947.1 | 16 | 1529 | ||
XM_006242738.2 | XP_006242800.1 | 15 | 1567 | |||
XM_008766062.1 | XP_008764284.1 | 17 | 2659 | |||
XM_008766063.1 | XP_008764285.1 | 16 | 1543 | |||
XM_008766064.1 | XP_008764286.1 | 15 | 1639 |