Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039477.2 | NP_001034566.1 | 4 | 1161 | ||
NM_001105556.2 | NP_001099026.1 | 6 | 2645 | |||
NM_001286113.1 | NP_001273042.1 | 7 | 2258 | |||
NM_001286115.1 | NP_001273044.1 | 6 | 2057 | |||
NM_004848.3 | NP_004839.2 | 5 | 1572 | |||
XM_005246041.3 | XP_005246098.1 | 5 | 2246 | |||
XM_006711050.1 | XP_006711113.1 | 7 | 2204 | |||
XM_011542445.2 | XP_011540747.1 | 5 | 2516 | |||
GenBank mRNA | AB050854.1 | - | 5 | 1861 | ||
AF044896.1 | - | 4 | 2298 | |||
AF323721.1 | - | 2 | 606 | |||
AK094833.1 | - | 2 | 2794 | |||
BC031655.1 | - | 5 | 1539 | |||
CR749321.1 | - | 4 | 1161 | |||
DB548309.2 | - | 1 | 597 |