Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033565.1 | NP_001028737.1 | 9 | 1337 | ||
XM_006233639.2 | XP_006233701.1 | 10 | 1609 | |||
XM_006233640.2 | XP_006233702.1 | 11 | 1703 | |||
XM_006233642.2 | XP_006233704.1 | 10 | 1594 | |||
XM_006233643.2 | XP_006233705.1 | 10 | 1579 | |||
GenBank mRNA | AY536920.1 | - | 9 | 1337 |