Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033565.1 | NP_001028737.1 | 4 | 415 | ||
XM_006233639.2 | XP_006233701.1 | 5 | 687 | |||
XM_006233640.2 | XP_006233702.1 | 6 | 781 | |||
XM_006233642.2 | XP_006233704.1 | 5 | 672 | |||
XM_006233643.2 | XP_006233705.1 | 5 | 687 | |||
GenBank mRNA | AY536920.1 | - | 4 | 415 |