Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pcp4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126093.2 | NP_001119565.1 | 3 | 271 | ||
XM_008769760.1 | XP_008767982.1 | 3 | 1112 | |||
GenBank mRNA | BC086330.1 | - | 3 | 271 | ||
BC162039.1 | - | 3 | 113 | 7099690 |