Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170326.1 | NP_001163797.1 | 11 | 2979 | ||
XM_008771857.1 | XP_008770079.1 | 9 | 1303 | |||
XM_008771858.1 | XP_008770080.1 | 8 | 1138 | |||
XM_008771859.1 | XP_008770081.1 | 9 | 1323 |