Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100909696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257589.2 | XP_006257651.1 | 1 | 105 |
Gene Symbol: LOC100364877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752159.3 | XP_003752207.1 | 1 | 105 |
Gene Symbol: LOC103694517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773575.1 | XP_008771797.1 | 1 | 248 |
Gene Symbol: LOC688702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257588.2 | XP_006257650.1 | 1 | 296 |
Gene Symbol: RGD1560585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257604.2 | XP_006257666.1 | 1 | 105 |
Gene Symbol: RGD1561558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228669.5 | XP_228669.3 | 105 |
Gene Symbol: RGD1566008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257584.2 | XP_006257646.1 | 1 | 290 |
Gene Symbol: RGD1566225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257564.2 | XP_006257626.1 | 1 | 105 |