Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 5 | 732 | ||
NM_001258401.2 | NP_001245330.1 | 5 | 678 | |||
NM_001258402.1 | NP_001245331.1 | 5 | 732 | |||
NM_001258403.1 | NP_001245332.1 | 5 | 804 | |||
NM_001267615.1 | NP_001254544.1 | 5 | 678 | |||
XM_006719700.1 | XP_006719763.1 | 5 | 733 | |||
XM_011539010.1 | XP_011537312.1 | 5 | 733 | |||
XM_011539011.1 | XP_011537313.1 | 5 | 678 | |||
XM_017020240.1 | XP_016875729.1 | 5 | 760 | |||
XM_017020241.1 | XP_016875730.1 | 5 | 1092 | |||
XM_017020242.1 | XP_016875731.1 | 5 | 747 | |||
GenBank mRNA | AB018282.1 | - | 7 | 812 | ||
AF069512.1 | - | 5 | 569 | |||
AF107099.1 | - | 5 | 568 | |||
AK128321.1 | - | 6 | 1232 | |||
AK295315.1 | - | 5 | 678 | |||
AK299545.1 | - | 2 | 364 | |||
AL831915.1 | - | 5 | 804 | |||
BC025994.1 | - | 5 | 620 | 4822306 | ||
DQ063579.2 | - | 5 | 721 | |||
DQ975204.1 | - | 5 | 548 | |||
DQ996398.1 | - | 5 | 782 | |||
DQ996537.1 | - | 5 | 548 |