Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014244.4 | NP_055059.2 | 9 | 1557 | ||
NM_021599.3 | NP_067610.1 | 9 | 1557 | |||
XM_017010063.1 | XP_016865552.1 | 8 | 993 | |||
GenBank mRNA | AJ003125.1 | - | 9 | 1455 | ||
AK308213.1 | - | 9 | 1560 |